1-bromo-4-iodonaphthalene ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
1-Bromo-4-iodonaphthalene
B4864-1G
B4864-5G
TCIAB4864-1GEA
411
DKK
TCIAB4864-1G
TCIAB4864-5G
1-bromo-4-iodonaphthalene ≥98.0% (by GC)
1-bromo-4-iodonaphthalene
Formel:
C₁₀H₆BrI Molekylvægt: 332.966 g/mol |
MDL Number:
MFCD01166242 CAS-nummer: 63279-58-3 |
Specifikationer Testresultater
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 88.0 to 92.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...