1,3-Butadien ∼13% ∼2 mol/L in THF

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Buta-1,3-diene

B4835-100ML B4835-500ML
TCIAB4835-100MLEA 304 DKK
TCIAB4835-100ML TCIAB4835-500ML
1,3-Butadien ∼13% ∼2 mol/L in THF
1,3-Butadiene
Formel: C₄H₆
Molekylvægt: 54.09 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00008659
CAS-nummer: 106-99-0
EINECS: 203-450-8
UN nummer: 1993
ADR: 3 II
Merck Index: 14,01509

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Concentration (HNMR) 13.0 to 15.0 w/w%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR