2-Brom-α,α,α,6-tetrafluortoluen ≥97.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
1-Bromo-3-fluoro-2-(trifluoromethyl)benzene, 2-Bromo-6-fluorobenzotrifluoride
B4833-1G
B4833-5G
TCIAB4833-1GEA
230
DKK
TCIAB4833-1G
TCIAB4833-5G
2-Brom-α,α,α,6-tetrafluortoluen ≥97.0% (by GC)
2-Brom-α,α,α,6-tetrafluortoluen
Formel:
C₇H₃BrF₄ |
MDL Number:
MFCD01631569 CAS-nummer: 261951-85-3 |
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...