5-Brom-o-toluonitril ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

B4808-1G B4808-5G
TCIAB4808-1GEA 345 DKK
TCIAB4808-1G TCIAB4808-5G
5-Brom-o-toluonitril ≥98.0% (by GC)
5-Brom-o-toluonitril
Formel: C₈H₆BrN
MDL Number: MFCD08061920
CAS-nummer: 156001-51-3
UN nummer: 3439
ADR: 6.1,III

Bestil nu

Specifikationer Testresultater

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 48.0 to 52.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR