O-(Benzotriazol-1-yl)-N,N,N',N'-bis(tetramethylene)uronium hexafluorophosphate ≥98.0% (by HPLC, total nitrogen)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: O-(Benzotriazol-1-yl)-N,N,N',N'-bis(tetramethylene)uronium hexafluorophosphate, (Benzotriazol-1-yloxy)dipyrrolidinocarbenium hexafluorophosphate, (1H-Benzo[d][1,2,3]triazol-1-yl)(di(pyrrolidin-1-yl)methylene)oxonium hexafluorophosphate(V)

B4805-1G B4805-5G
TCIAB4805-1GEA 485 DKK
TCIAB4805-1G TCIAB4805-5G
O-(Benzotriazol-1-yl)-N,N,N',N'-bis(tetramethylene)uronium hexafluorophosphate ≥98.0% (by HPLC, total nitrogen)
O-(Benzotriazol-1-yl)-N,N,N',N'-bis(tetramethylene)uronium hexafluorophosphate
Formel: C₁₅H₂₀F₆N₅OP
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00191770
CAS-nummer: 105379-24-6

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR