2-Bromo-4'-methylpropiophenone ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
B4781-5G
B4781-25G
TCIAB4781-5GEA
214
DKK
TCIAB4781-5G
TCIAB4781-25G
2-Bromo-4'-methylpropiophenone ≥98.0% (by GC)
2-Bromo-4'-methylpropiophenone
Formel:
C₁₀H₁₁BrO Molekylvægt: 227.101 g/mol Kogepunkt: 162°C |
MDL Number:
MFCD11131402 CAS-nummer: 1451-82-7 |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 78.0 to 82.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...