5-Bromo-1-methylisatin ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
5-Bromo-1-methylindoline-2,3-dione
B4779-1G
B4779-5G
TCIAB4779-1GEA
591
DKK
TCIAB4779-1G
TCIAB4779-5G
5-Bromo-1-methylisatin ≥98.0% (by GC)
5-Bromo-1-methylisatin
Formel:
C₉H₆BrNO₂ Molekylvægt: 240.06 g/mol |
MDL Number:
MFCD00456313 CAS-nummer: 2058-72-2 |
Specifikationer Testresultater
Appearance | Light red to Yellow to Red powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 169.0 to 173.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...