2-(Benzyloxy)benzoesyre ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-Benzyloxybenzoic acid

B4766-5G B4766-25G
TCIAB4766-5GEA 493 DKK
TCIAB4766-5G TCIAB4766-25G
2-(Benzyloxy)benzoesyre ≥98.0% (by GC)
2-(Benzyloxy)benzoesyre
Formel: C₁₄H₁₂O₃
Molekylvægt: 228.25 g/mol
MDL Number: MFCD00051940
CAS-nummer: 14389-86-7
UN nummer: 3077
ADR: 9 III

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 95.0 %
Melting point 74.0 to 78.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR