4-Bromocarbazole ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
4-Bromo-9H-carbazole
B4752-200MG
B4752-1G
TCIAB4752-200MGEA
173
DKK
TCIAB4752-200MG
TCIAB4752-1G
4-Bromocarbazole ≥98.0% (by GC)
4-Bromocarbazole
Formel:
C₁₂H₈BrN |
MDL Number:
MFCD18450175 CAS-nummer: 3652-89-9 |
Specifikationer Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 105.0 to 109.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...