5-Brom-2-methoxybenzoesyre ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

B4741-1G B4741-5G
TCIAB4741-1GEA 197 DKK
TCIAB4741-1G TCIAB4741-5G
5-Brom-2-methoxybenzoesyre ≥98.0% (by GC, titration analysis)
5-Brom-2-methoxybenzoesyre
Formel: C₈H₇BrO₃
Molekylvægt: 231.05 g/mol
MDL Number: MFCD00093713
CAS-nummer: 2476-35-9

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 119.0 to 123.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR