3-Bromo-5-(trifluoromethyl)pyridine ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
B4710-1G
B4710-5G
TCIAB4710-1GEA
796
DKK
TCIAB4710-1G
TCIAB4710-5G
3-Bromo-5-(trifluoromethyl)pyridine ≥98.0% (by GC, titration analysis)
3-Bromo-5-(trifluoromethyl)pyridine
Formel:
C₆H₃BrF₃N Kogepunkt: 66°C Flammepunkt: 85°C |
MDL Number:
MFCD04972700 CAS-nummer: 436799-33-6 UN nummer: 2811 ADR: 6.1,III |
|
Specifikationer Testresultater
Appearance | White to Almost white powder to lump |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 42.0 to 46.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...