Benzyl-4-chlorophenyl ketone ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Benzyl 4-chlorophenyl ketone, 4'-Chloro-2-phenylacetophenone

B4704-5G B4704-25G
TCIAB4704-5GEA 279 DKK
TCIAB4704-5G TCIAB4704-25G
Benzyl-4-chlorophenyl ketone ≥98.0% (by GC)
Benzyl-4-chlorophenyl ketone
Formel: C₁₄H₁₁ClO
Molekylvægt: 230.69 g/mol
Kogepunkt: 187°C
MDL Number: MFCD00016342
CAS-nummer: 1889-71-0

Bestil nu

Specifikationer Testresultater

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 104.0 to 108.0 deg-C

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR