4-Brom-3-(trifluormethyl)benzonitril ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
B4691-1G
B4691-5G
TCIAB4691-1GEA
362
DKK
TCIAB4691-1G
TCIAB4691-5G
4-Brom-3-(trifluormethyl)benzonitril ≥98.0% (by GC)
4-Brom-3-(trifluormethyl)benzonitril
Formel:
C₈H₃BrF₃N Molekylvægt: 250.02 g/mol |
MDL Number:
MFCD03095347 CAS-nummer: 1735-53-1 UN nummer: 3439 ADR: 6.1 III |
Specifikationer Testresultater
Appearance | Light orange to Yellow to Green powder to lump |
Purity (GC) | min. 98.0 % |
Melting point | 80.0 to 84.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...