4-Bromo-1H-indazole ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

B4631-1G B4631-5G
TCIAB4631-1GEA 271 DKK
TCIAB4631-1G TCIAB4631-5G
4-Bromo-1H-indazole ≥98.0% (by GC)
4-Bromo-1H-indazole
Formel: C₇H₅BrN₂
MDL Number: MFCD05664001
CAS-nummer: 186407-74-9

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 165.0 to 169.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR