Boc-Asp-OtBu ≥95.0% (by HPLC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
B4618-1G
B4618-5G
TCIAB4618-1GEA
312
DKK
TCIAB4618-1G
TCIAB4618-5G
Boc-Asp-OtBu ≥95.0% (by HPLC, titration analysis)
Boc-Asp-OtBu
Formel:
C₁₃H₂₃NO₆ Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD00038272 CAS-nummer: 34582-32-6 |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 95.0 % (Exclude toluene) |
Purity (Neutralization titration) | min. 95.0 % |
Melting point | 101.0 to 106.0 deg-C |
Specific rotation [a]20/D | -22.0 to -25.0 deg (C=1.5, MeOH) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...