2-Brom-4-thiazolcarboxylsyre ≥98.0% (by titrimetric analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
2-Bromo-1,3-thiazole-4-carboxylic acid, 2-Bromothiazole-4-carboxylic acid
B4578-1G
B4578-5G
TCIAB4578-1GEA
362
DKK
TCIAB4578-1G
TCIAB4578-5G
2-Brom-4-thiazolcarboxylsyre ≥98.0% (by titrimetric analysis)
2-Brom-4-thiazolcarboxylsyre
Formel:
C₄H₂BrNO₂S Smeltepunkt: 217°C |
MDL Number:
MFCD04115729 CAS-nummer: 5198-88-9 |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...