2-Bromfuran ≥98.0% (by GC) stabilised
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Stabilization:
Stabilised with calcia (CaO)
B4555-1G
B4555-5G
TCIAB4555-1GEA
321
DKK
TCIAB4555-1G
TCIAB4555-5G
2-Bromfuran ≥98.0% (by GC) stabilised
2-Bromofuran
Formel:
C₄H₃BrO Kogepunkt: 102°C Opbevaringstemperatur: Fryser |
MDL Number:
MFCD00216609 CAS-nummer: 584-12-3 UN nummer: 1993 ADR: 3,II |
Specifikationer Testresultater
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...