7-Brom-1-tetralon ≥95.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 7-Bromo-3,4-dihydronaphthalen-1(2H)-one

B4545-1G B4545-5G
TCIAB4545-1GEA 312 DKK
TCIAB4545-1G TCIAB4545-5G
7-Brom-1-tetralon ≥95.0% (by GC)
7-Brom-1-tetralon
Formel: C₁₀H₉BrO
Molekylvægt: 225.08 g/mol
Kogepunkt: 157°C
MDL Number: MFCD02179287
CAS-nummer: 32281-97-3

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow to Dark green powder to crystal
Purity (GC) min. 95.0 %
Melting point 74.0 to 78.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR