Boc-Glu-OtBu ≥97.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: N-(tert-Butoxycarbonyl)-L-glutamic Acid 1-tert-Butyl Ester, N-Boc-L-glutamic Acid 1-tert-Butyl Ester, 1-tert-Butyl N-Boc-L-glutamate, Boc-Glu-OtBu

B4534-5G B4534-25G
TCIAB4534-5GEA 542 DKK
TCIAB4534-5G TCIAB4534-25G
Boc-Glu-OtBu ≥97.0% (by HPLC, titration analysis)
Boc-Glu-OtBu
Formel: C₁₄H₂₅NO₆
Opbevaringstemperatur: Fryser
MDL Number: MFCD00038273
CAS-nummer: 24277-39-2

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystaline
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) min. 97.0 %
Melting point 111.0 to 115.0 deg-C
Specific rotation [a]20/D -32.0 to -37.0 deg (C=0.6,MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR