1,3-Bis(4-fluorophenyl)urea ≥98.0% (by HPLC, total nitrogen)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

B4517-1G B4517-5G
TCIAB4517-1GEA 608 DKK
TCIAB4517-1G TCIAB4517-5G
1,3-Bis(4-fluorophenyl)urea ≥98.0% (by HPLC, total nitrogen)
1,3-Bis(4-fluorophenyl)urea
Formel: C₁₃H₁₀F₂N₂O
MDL Number: MFCD00017925
CAS-nummer: 370-22-9

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR