6-Brom-m-toluidin ≥97.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
B4507-5G
B4507-25G
TCIAB4507-5GEA
247
DKK
TCIAB4507-5G
TCIAB4507-25G
6-Brom-m-toluidin ≥97.0% (by GC, titration analysis)
6-Brom-m-toluidin
Formel:
C₇H₈BrN Molekylvægt: 186.05 g/mol Kogepunkt: 130°C Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD01806278 CAS-nummer: 53078-85-6 |
Specifikationer Testresultater
Appearance | White to Orange to Green powder to lump |
Purity (GC) | min. 97.0 % |
Purity (Nonaqueous Titration) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...