4-Bromo-3-(trifluoromethyl)phenol ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-Bromo-5-hydroxybenzotrifluoride

B4494-1G B4494-5G
TCIAB4494-1GEA 132 DKK
TCIAB4494-1G TCIAB4494-5G
4-Bromo-3-(trifluoromethyl)phenol ≥98.0% (by GC)
4-Bromo-3-(trifluoromethyl)phenol
Formel: C₇H₄BrF₃O
Molekylvægt: 241.01 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD03095342
CAS-nummer: 320-49-0

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 46.0 to 50.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR