N-(tert-Butoxycarbonyl)propargylamine ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: N-Boc-propargylamine, 2-Propynylcarbamic Acid tert-Butyl Ester, tert-Butyl 2-Propynylcarbamate

B4472-1G B4472-5G
TCIAB4472-1GEA 132 DKK
TCIAB4472-1G TCIAB4472-5G
N-(tert-Butoxycarbonyl)propargylamine ≥98.0% (by GC)
N-(tert-Butoxycarbonyl)propargylamine
Formel: C₈H₁₃NO₂
Kogepunkt: 170°C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD07367245
CAS-nummer: 92136-39-5

Bestil nu

Specifikationer Testresultater

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 40.0 to 44.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR