3-(2-Bromphenyl)propionsyre ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
B4409-5G
B4409-25G
TCIAB4409-5GEA
230
DKK
TCIAB4409-5G
TCIAB4409-25G
3-(2-Bromphenyl)propionsyre ≥98.0% (by GC, titration analysis)
3-(2-Bromphenyl)propionsyre
Formel:
C₉H₉BrO₂ Molekylvægt: 229.07 g/mol Kogepunkt: 186°C |
MDL Number:
MFCD01310791 CAS-nummer: 15115-58-9 |
Specifikationer Testresultater
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 97.0 to 101.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...