Bilobalide ≥96.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

B4388-10MG B4388-50MG
TCIAB4388-10MGEA 550 DKK
TCIAB4388-10MG TCIAB4388-50MG
Bilobalide ≥96.0% (by GC)
Bilobalide
Formel: C₁₅H₁₈O₈
Opbevaringstemperatur: Fryser
MDL Number: MFCD00238547
CAS-nummer: 33570-04-6
Merck Index: 14,01220

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 96.0 %
Specific rotation [a]20/D -57.0 to -63.0 deg (C=1, Acetone)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR