(4-Bromphenyl)acetone ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 4-Bromophenylacetone

B4387-5G B4387-25G
TCIAB4387-5GEA 419 DKK
TCIAB4387-5G TCIAB4387-25G
(4-Bromphenyl)acetone ≥98.0% (by GC)
(4-Bromphenyl)acetone
Formel: C₉H₉BrO
Kogepunkt: 139°C
MDL Number: MFCD00210401
CAS-nummer: 6186-22-7

Bestil nu

Specifikationer Testresultater

Appearance Light orange to Yellow to Green clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR