tert-Butyl-3-(isopropyl)carbazate ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 1-tert-Butoxycarbonyl-2-isopropylhydrazine, 3-(Isopropyl)carbazic Acid tert-Butyl Ester, 1-Boc-2-isopropylhydrazine, tert-Butyl 2-isopropylhydrazinecarboxylate

B4375-1G B4375-5G
TCIAB4375-1GEA 378 DKK
TCIAB4375-1G TCIAB4375-5G
tert-Butyl-3-(isopropyl)carbazate ≥98.0% (by GC, titration analysis)
tert-Butyl-3-(isopropyl)carbazate
Formel: C₈H₁₈N₂O₂
Molekylvægt: 174.24 g/mol
Kogepunkt: 80°C
MDL Number: MFCD14635748
CAS-nummer: 16689-35-3

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 52.0 to 56.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR