Balofloxacin dihydrate ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 1-Cyclopropyl-6-fluoro-1,4-dihydro-8-methoxy-7-[3-(methylamino)-1-piperidyl]-4-oxoquinoline-3-carboxylic acid

B4354-1G B4354-5G
TCIAB4354-1GEA 444 DKK
TCIAB4354-1G TCIAB4354-5G
Balofloxacin dihydrate ≥98.0% (by HPLC, titration analysis)
Balofloxacin dihydrate
Formel: C₂₀H₂₄FN₃O₄·2H₂O
Smeltepunkt: 135°C
MDL Number: MFCD00864925
CAS-nummer: 151060-21-8

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 % (as Dihydrate)
Water 6.5 to 10.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR