6-Bromo-1H-indole-2-carboxylic acid ≥97.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
6-Bromoindole-2-carboxylic acid
B4334-1G
B4334-5G
TCIAB4334-1GEA
534
DKK
TCIAB4334-1G
TCIAB4334-5G
6-Bromo-1H-indole-2-carboxylic acid ≥97.0% (by GC)
6-Bromo-1H-indole-2-carboxylic acid
Formel:
C₉H₆BrNO₂ Molekylvægt: 240.05 g/mol |
MDL Number:
MFCD02664469 CAS-nummer: 16732-65-3 |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (Neutralization titration) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...