2-Brom-5-thiazolcarboxylsyre ≥97.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
2-Bromothiazole-5-carboxylic acid
B4329-1G
B4329-5G
TCIAB4329-1GEA
353
DKK
TCIAB4329-1G
TCIAB4329-5G
2-Brom-5-thiazolcarboxylsyre ≥97.0% (by GC, titration analysis)
2-Brom-5-thiazolcarboxylsyre
Formel:
C₄H₂BrNO₂S |
MDL Number:
MFCD04115730 CAS-nummer: 54045-76-0 |
Specifikationer Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (Neutralization titration) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...