2-Bromo-9,10-bis(2-naphthalenyl)anthracene ≥98.0% (by HPLC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 2-Bromo-9,10-di(2-naphthyl)anthracene

B4321-1G B4321-5G
TCIAB4321-1GEA 263 DKK
TCIAB4321-1G TCIAB4321-5G
2-Bromo-9,10-bis(2-naphthalenyl)anthracene ≥98.0% (by HPLC)
2-Bromo-9,10-bis(2-naphthalenyl)anthracene
Formel: C₃₄H₂₁Br
MDL Number: MFCD11045064
CAS-nummer: 474688-76-1

Bestil nu

Specifikationer Testresultater

Appearance White to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR