2-Brom-5-chlor-3-nitropyridin ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

B4310-5G B4310-25G
TCIAB4310-5GEA 345 DKK
TCIAB4310-5G TCIAB4310-25G
2-Brom-5-chlor-3-nitropyridin ≥98.0% (by GC)
2-Bromo-5-chloro-3-nitropyridine
Formel: C₅H₂BrClN₂O₂
MDL Number: MFCD07368982
CAS-nummer: 75806-86-9

Bestil nu

Specifikationer Testresultater

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 75.0 to 79.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR