1,3,5-Benzenetricarbonitrile ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

B4294-1G B4294-5G
TCIAB4294-1GEA 903 DKK
TCIAB4294-1G TCIAB4294-5G
1,3,5-Benzenetricarbonitrile ≥98.0% (by GC)
1,3,5-Benzenetricarbonitrile
Formel: C₉H₃N₃
MDL Number: MFCD00019746
CAS-nummer: 10365-94-3
EINECS: 233-806-8
UN nummer: 3439
ADR: 6.1,III

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR