N-(tert-Butoxycarbonyl)-4-aminobutyric acid ≥98.0% (by titrimetric analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
Boc-GABA-OH, N-Boc-γ-aminobutyric acid, 4-(Boc-amino)butyric Acid, 4-(tert-Butoxycarbonylamino)butyric Acid, N-Boc-4-aminobutyric Acid, N-BOC-gamma-Aminobutyric acid
B4291-5G
B4291-25G
TCIAB4291-5GEA
156
DKK
TCIAB4291-5G
TCIAB4291-25G
N-(tert-Butoxycarbonyl)-4-aminobutyric acid ≥98.0% (by titrimetric analysis)
N-(tert-Butoxycarbonyl)-4-aminobutyric acid
Formel:
C₉H₁₇NO₄ Kogepunkt: 82°C |
MDL Number:
MFCD00037313 CAS-nummer: 57294-38-9 |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 95.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 56.0 to 60.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...