N-(tert-Butoxycarbonyl)-L-methioninol ≥95.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: (S)-2-(tert-Butoxycarbonyl)amino-4-methylthio-1-butanol, Boc-Methioninol, N-Boc-L-methioninol, (S)-2-(Boc-amino)-4-methylthio-1-butanol, (S)-tert-Butyl (1-hydroxy-4-(methylthio)butan-2-yl)carbamate

B4283-5G B4283-1G
TCIAB4283-5GEA 2320 DKK
TCIAB4283-5G TCIAB4283-1G
N-(tert-Butoxycarbonyl)-L-methioninol ≥95.0% (by GC)
N-(tert-Butoxycarbonyl)-L-methioninol
Formel: C₁₀H₂₁NO₃S
MDL Number: MFCD00235939
CAS-nummer: 51372-93-1

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 95.0 %
Melting point 48.0 to 52.0 deg-C
Specific rotation [a]20/D -18.0 to -21.0 deg (C=1, MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR