2,2'-Bipyridine-3,3'-diol ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
3,3'-Dihydroxy-2,2'-bipyridine, 3,3'-Dihydroxy-2,2'-bipyridyl
B4255-50MG
B4255-250MG
TCIAB4255-50MGEA
1120
DKK
TCIAB4255-50MG
TCIAB4255-250MG
2,2'-Bipyridine-3,3'-diol ≥98.0% (by GC)
2,2'-Bipyridine-3,3'-diol
Formel:
C₁₀H₈N₂O₂ |
MDL Number:
MFCD00075177 CAS-nummer: 36145-03-6 |
Specifikationer Testresultater
Purity (GC) | min. 98.0 % |
Melting point | 197.0 to 201.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...