1-Brom-5-Chlor-2-iodbenzen ≥96.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

B4243-5G B4243-25G
TCIAB4243-5GEA 279 DKK
TCIAB4243-5G TCIAB4243-25G
1-Brom-5-Chlor-2-iodbenzen ≥96.0% (by GC)
1-Brom-5-Chlor-2-iodbenzen
Formel: C₆H₃BrClI
Molekylvægt: 317.35 g/mol
Kogepunkt: 111°C
Smeltepunkt: 33°C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00672944
CAS-nummer: 31928-44-6

Bestil nu

Specifikationer Testresultater

Appearance White or Colorless to Yellow powder to lump to clear liquid
Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR