1-Butyl-2,3-dimethylimidazolium trifluormethansulphonat ≥98.0% (by HPLC, total nitrogen)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 1-Butyl-2,3-dimethylimidazolium triflate, 1-Butyl-2,3-dimethylimidazolium Trifluoromethanesulphonate

B4182-5G B4182-25G
TCIAB4182-5GEA 624 DKK
TCIAB4182-5G TCIAB4182-25G
1-Butyl-2,3-dimethylimidazolium trifluormethansulphonat ≥98.0% (by HPLC, total nitrogen)
1-Butyl-2,3-dimethylimidazolium trifluormethansulphonat
Formel: C₁₀H₁₇F₃N₂O₃S
Molekylvægt: 302.32 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD03427624
CAS-nummer: 765910-73-4

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 42.0 to 46.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR