4,4'-Biphenyldiboronic acid bis(pinacol) ester ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
4,4,5,5-Tetramethyl-2-{4-[4-(tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]phenyl}-1,3,2-dioxaborolane, 4,4'-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,1'-biphenyl
B4166-5G
B4166-1G
TCIAB4166-5GEA
2640
DKK
TCIAB4166-5G
TCIAB4166-1G
4,4'-Biphenyldiboronic acid bis(pinacol) ester ≥98.0% (by GC, titration analysis)
4,4'-Biphenyldiboronic acid bis(pinacol)ester
Formel:
C₂₄H₃₂B₂O₄ Molekylvægt: 406.14 g/mol Smeltepunkt: 262°C |
MDL Number:
MFCD08669545 CAS-nummer: 207611-87-8 |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...