brompentamethylbenzen ≥97.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 1-Bromo-2,3,4,5,6-pentamethylbenzene, Pentamethylbromobenzene

B4154-5G B4154-25G
TCIAB4154-5GEA 304 DKK
TCIAB4154-5G TCIAB4154-25G
brompentamethylbenzen ≥97.0% (by GC)
brompentamethylbenzen
Formel: C₁₁H₁₅Br
Kogepunkt: 290°C
MDL Number: MFCD00013523
CAS-nummer: 5153-40-2
EINECS: 225-926-4

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Melting point 160.0 to 164.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR