Balsalazide disodium salt dihydrate ≥98.0% (by HPLC, total nitrogen)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 5-[4-(2-Carboxyethylcarbamoyl)phenylazo]salicylic Acid Disodium Salt, Sodium (E)-5-((4-((2-carboxylatoethyl)carbamoyl)phenyl)diazenyl)-2-hydroxybenzoate dihydrate

B4121-1G B4121-5G
TCIAB4121-1GEA 419 DKK
TCIAB4121-1G TCIAB4121-5G
Balsalazide disodium salt dihydrate ≥98.0% (by HPLC, total nitrogen)
Balsalazide disodium salt dihydrate
Formel: C₁₇H₁₇N₃Na₂O₈
MDL Number: MFCD01631117
CAS-nummer: 150399-21-6
Merck Index: 14,00945

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 % (calcd.on anh.substance)
Water 7.0 to 11.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR