Biochanin A ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
5,7-Dihydroxy-3-(4-methoxyphenyl)-4-benzopyrone, 5,7-Dihydroxy-3-(4-methoxyphenyl)chromen-4-one, 4'-Methoxy-5,7-dihydroxyisoflavone, 5,7-Dihydroxy-3-(4-methoxyphenyl)-4H-1-benzopyran-4-one, 5,7-Dihydroxy-3-(4-methoxyphenyl)-4H-chromen-4-one, 5,7-Dihydroxy-4'-methoxyisoflavone
B4098-200MG
B4098-1G
B4098-5G
TCIAB4098-200MGEA
279
DKK
TCIAB4098-200MG
TCIAB4098-1G
TCIAB4098-5G
Biochanin A ≥98.0% (by HPLC, titration analysis)
Biochanin A
Formel:
C₁₆H₁₂O₅ |
MDL Number:
MFCD00006839 CAS-nummer: 491-80-5 EINECS: 207-744-7 |
Specifikationer Testresultater
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 215.0 to 219.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...