3-tert-Butylanilin ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
3-(tert-Butyl)aniline, m-tert-Butylaniline
B4072-1G
B4072-5G
TCIAB4072-1GEA
690
DKK
TCIAB4072-1G
TCIAB4072-5G
3-tert-Butylanilin ≥98.0% (by GC, titration analysis)
3-tert-Butylanilin
Formel:
C₁₀H₁₅N Molekylvægt: 149.24 g/mol Kogepunkt: 119°C |
MDL Number:
MFCD00125078 CAS-nummer: 5369-19-7 EINECS: 226-361-6 |
Specifikationer Testresultater
Appearance | Light orange to Yellow to Green clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...