N,N-Bis(2-chloroethyl)-p-toluenesulphonamide ≥98.0% (by HPLC, total nitrogen)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: N,N-Bis(2-chloroethyl)-4-methylbenzenesulphonamide

B4059-5G B4059-25G
TCIAB4059-5GEA 403 DKK
TCIAB4059-5G TCIAB4059-25G
N,N-Bis(2-chloroethyl)-p-toluenesulphonamide ≥98.0% (by HPLC, total nitrogen)
N,N-Bis(2-chloroethyl)-p-toluenesulphonamide
Formel: C₁₁H₁₅Cl₂NO₂S
Molekylvægt: 296.21 g/mol
MDL Number: MFCD00018944
CAS-nummer: 42137-88-2
EINECS: 255-677-7

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 45.0 to 49.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR