2-(Bromomethyl)acrylic acid ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

B4048-1G B4048-5G
TCIAB4048-1GEA 895 DKK
TCIAB4048-1G TCIAB4048-5G
2-(Bromomethyl)acrylic acid ≥98.0% (by GC, titration analysis)
2-(Bromomethyl)acrylic acid
Formel: C₄H₅BrO₂
Molekylvægt: 164.99 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00004259
CAS-nummer: 72707-66-5
EINECS: 276-774-0
UN nummer: 3261
ADR: 8 II

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 71.0 to 75.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR