5-tert-Butyl-L-glutamate hydrate ≥98.0% (by titrimetric analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
H-Glu(OtBu)-OH, L-Glutamic Acid 5-tert-Butyl Ester
B4045-1G
B4045-5G
TCIAB4045-1GEA
197
DKK
TCIAB4045-1G
TCIAB4045-5G
5-tert-Butyl-L-glutamate hydrate ≥98.0% (by titrimetric analysis)
5-tert-Butyl-L-glutamate hydrate
Formel:
C₉H₁₇NO₄ Smeltepunkt: 182°C Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD00038580 CAS-nummer: 2419-56-9 |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % (calcd.on anh.substance) |
Specific rotation [a]20/D | +9.0 to +12.0 deg (C=1、H2O) (calcd.on anh.substance) |
Water | 3.0 to 6.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...