1,4-Bis[(1H-imidazol-1-yl)methyl]benzene ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

B4023-5G B4023-1G
TCIAB4023-5GEA 1600 DKK
TCIAB4023-5G TCIAB4023-1G
1,4-Bis[(1H-imidazol-1-yl)methyl]benzene ≥98.0% (by GC)
1,4-Bis[(1H-imidazol-1-yl)methyl]benzene
Formel: C₁₄H₁₄N₄
MDL Number: MFCD06598031
CAS-nummer: 56643-83-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d25341bc70a8076b1d319b5876dead9ea43fe956&vid=d25341bc70a8076b1d319b5876dead9ea43fe956&prid=web&clid=SAPDAM

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR