5-Benzyl-D-glutamate ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
D-Glutamic Acid 5-Benzyl Ester, (R)-2-Amino-5-(benzyloxy)-5-oxopentanoic acid, H-D-Glu(OBzl)-OH
B3999-1G
B3999-5G
TCIAB3999-1GEA
132
DKK
TCIAB3999-1G
TCIAB3999-5G
5-Benzyl-D-glutamate ≥98.0% (by HPLC, titration analysis)
5-Benzyl-D-glutamate
Formel:
C₁₂H₁₅NO₄ Molekylvægt: 237.26 g/mol |
MDL Number:
MFCD00063111 CAS-nummer: 2578-33-8 |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 157.0 to 161.0 deg-C |
Specific rotation [a]20/D | -17.0 to -21.0 deg (C=1、AcOH) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...