1-Boc-1-methylhydrazine ≥97.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
B3998-5G
B3998-25G
TCIAB3998-5GEA
255
DKK
TCIAB3998-5G
TCIAB3998-25G
1-Boc-1-methylhydrazine ≥97.0% (by GC, titration analysis)
1-Boc-1-methylhydrazine
Formel:
C₆H₁₄N₂O₂ Kogepunkt: 80°C Flammepunkt: 71°C |
MDL Number:
MFCD05669700 CAS-nummer: 21075-83-2 |
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Purity (Nonaqueous Titration) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...