(R)-2-((tert-Butoxycarbonyl)amino)-3,3-dimethylbutanoic acid ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Boc-D-Tle-OH, N-Boc-D-tert-leucine, Boc-D-Tle-O, (R)-2-(tert-Butoxycarbonylamino)-3,3-dimethylbutyric Acid, (R)-2-(Boc-amino)-3,3-dimethylbutyric Acid, Boc-tBu-D-Gly-OH

B3995-5G B3995-1G
TCIAB3995-5GEA 1340 DKK
TCIAB3995-5G TCIAB3995-1G
(R)-2-((tert-Butoxycarbonyl)amino)-3,3-dimethylbutanoic acid ≥98.0% (by HPLC, titration analysis)
(R)-2-((tert-Butoxycarbonyl)amino)-3,3-dimethylbutanoic acid
Formel: C₁₁H₂₁NO₄
Molekylvægt: 231.29 g/mol
MDL Number: MFCD00065575
CAS-nummer: 124655-17-0

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 117.0 to 122.0 deg-C
Specific rotation [a]20/D +4.0 to +7.0 deg (C=1、AcOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR